1,4-Di(chlormethyl)-2,3,5,6-tetramethylbenzol

Lieferant: CHEMPUR
Gefahr

Synonyme: 1,4-Bis(chloromethyl)-2,3,5,6-tetramethylbenzene, 3,6-Bis(chloromethyl)durene

BTB14145.250 BTB14145.1
CHMPBTB14145.250EA 0 EUR
CHMPBTB14145.250 MAYBBTB14145.1
1,4-Di(chlormethyl)-2,3,5,6-tetramethylbenzol
1,4-Di(chlormethyl)-2,3,5,6-tetramethylbenzol
Formel: C₁₂H₁₆Cl₂
Molecular Weight: 231.16 g/mol
MDL: MFCD00018883
CAS-Nummer: 3022-16-0
EINECS: 221-173-0
UN: 3335
ADR: 9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR