Hexamethylbenzol 99

Lieferant: Thermo Fisher Scientific

A10294.03 A10294.06 A10294.14
A10294.03EA 31.5 EUR
A10294.03 A10294.06 A10294.14
Hexamethylbenzol 99
Hexamethylbenzene

Purity: 99+% CAS No.: 87-85-4 Molecular Formula: C12H18 Formula Weight: 162.28 Melting Point: 163-167° Boiling Point: 265° Solubility: Insoluble in water Beilstein Reference: 1905834 MDL No.: MFCD00008523

Formel: C₁₂H₁₈
Molecular Weight: 162.27 g/mol
Siedepunkt: 265 °C
Schmelzpunkt: 163…167 °C
Dichte: 1.063
Lagertemperatur: Raumtemperatur
MDL: MFCD00008523
CAS-Nummer: 87-85-4
EINECS: 201-777-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR