Cyclohexen-1-ylbenzol ≥97%
Lieferant: Thermo Scientific
Synonyme:
1-Phenylcyclohexen
B20602.06
B20602.14
B20602.22
B20602.06EA
45.2
EUR
B20602.06
B20602.14
B20602.22
Cyclohexen-1-ylbenzol ≥97%
Cyclohexen-1-ylbenzol
Purity: 97%
CAS No.: 771-98-2
Molecular Formula: C12H14
Formula Weight: 158.24
Flash Point: 103°(217°F)
Refractive Index: 1.5700
Melting Point: -11°
Boiling Point: 250-252°
Beilstein Reference: 1905772
MDL No.: MFCD00001542
Formel:
C₁₂H₁₄ Molecular Weight: 158,24 g/mol Siedepunkt: 250…252 °C Schmelzpunkt: –11 °C Dichte: 0,992 Flammpunkt: 103 °C (217 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001542 CAS-Nummer: 771-98-2 EINECS: 212-242-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...