3-(Benzyloxy)benzaldehyd ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 3-Benzyloxybenzaldehyde

B22909.06 B22909.14 B22909.22
B22909.06EA 27.1 EUR
B22909.06 B22909.14 B22909.22
3-(Benzyloxy)benzaldehyd ≥98%
3-(Benzyloxy)benzaldehyd

Purity: 98% CAS No.: 1700-37-4 Molecular Formula: C14H12O2 Formula Weight: 212.25 Melting Point: 54-58° Boiling Point: 180°/0.08mm MDL No.: MFCD00003367

Formel: C₁₄H₁₂O₂
Molecular Weight: 212.25 g/mol
Siedepunkt: 180 °C (0.08 mmHg)
Schmelzpunkt: 54…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003367
CAS-Nummer: 1700-37-4
EINECS: 216-932-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR