2-Phenylbenzoesäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Biphenyl-2-carbonsäure, 2-Biphenylcarbonsäure

A12049.06 A12049.14 A12049.22
A12049.06EA 21.7 EUR
A12049.06 A12049.14 A12049.22
2-Phenylbenzoesäure ≥98%
2-Phenylbenzoesäure

Purity: 98% CAS No.: 947-84-2 Molecular Formula: C13H10O2 Formula Weight: 198.22 Odour: Odourless Melting Point: 110-114° Boiling Point: 343-344° Beilstein Reference: 974075 MDL No.: MFCD00002463

Formel: C₁₃H₁₀O₂
Molecular Weight: 198,22 g/mol
Siedepunkt: 343…344 °C
Schmelzpunkt: 110…114 °C
Dichte: 1,458 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002463
CAS-Nummer: 947-84-2
EINECS: 213-432-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR