Bicyclo[3.2.1]-2-octanon

Lieferant: CHEMPUR

BTB12718.250 BTB12718.1
CHMPBTB12718.250EA 0 EUR
CHMPBTB12718.250 MAYBBTB12718.1
Bicyclo[3.2.1]-2-octanon
Bicyclo[3.2.1]-2-octanon
Formel: C₈H₁₂O
Molecular Weight: 124,18 g/mol
Siedepunkt: 85…110 °C (21 mmHg)
Schmelzpunkt: 144…145 °C
CAS-Nummer: 5019-82-9
EINECS: 225-704-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR