5-Methylbicyclo[2.2.1]-2-hepten

Lieferant: Thermo Scientific

BTB10733.250 BTB10733.1
MAYBBTB10733.250EA 0 EUR
MAYBBTB10733.250 MAYBBTB10733.1
5-Methylbicyclo[2.2.1]-2-hepten
5-Methylbicyclo[2.2.1]-2-hepten
Formel: C₈H₁₂
Molecular Weight: 108.18 g/mol
CAS-Nummer: 822-96-8
EINECS: 212-509-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR