5-Bromchinolin ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
L19634.03
L19634.MD
L19634.14
L19634.06
L19634.03EA
25.2
EUR
L19634.03
L19634.MD
L19634.14
L19634.06
5-Bromchinolin ≥97%
5-Bromoquinoline
Purity: 97%
CAS No.: 4964-71-0
Molecular Formula: C9H6BrN
Formula Weight: 208.06
Melting Point: 44-48°
Boiling Point: 105-107°/1mm
MDL No.: MFCD00234481
Formel:
C₉H₆BrN Molecular Weight: 208,06 g/mol Siedepunkt: 105…107 °C (1 mmHg) Schmelzpunkt: 44…48 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234481 CAS-Nummer: 4964-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...