2-Chinolincarbaldehyd 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Chinolincarboxaldehyd
370170050
370170250
ACRO370170050EA
71.1
EUR
ACRO370170050
ACRO370170250
2-Chinolincarbaldehyd 97%
2-Chinolincarbaldehyd
Formel:
C₁₀H₇NO Molecular Weight: 157,17 g/mol Schmelzpunkt: 66…71 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00075032 CAS-Nummer: 5470-96-2 EINECS: 226-804-3 |
Spezifikation Testergebnisse
Appearance | Yellow to orange to brown Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 66 to 71 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...