2-Chinolincarbaldehyd ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Chinolincarboxaldehyd
L20202.06
L20202.14
L20202.06EA
71.4
EUR
L20202.06
L20202.14
2-Chinolincarbaldehyd ≥97%
2-Chinolincarbaldehyd
Purity: 97%
CAS No.: 5470-96-2
Molecular Formula: C10H7NO
Formula Weight: 157.17
Melting Point: 66-70°
Beilstein Reference: 113061
Formel:
C₁₀H₇NO Molecular Weight: 157.17 g/mol Schmelzpunkt: 66…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075032 CAS-Nummer: 5470-96-2 EINECS: 226-804-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...