5-Chinolincarbaldehyd ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Chinolincarboxaldehyd

L19641.03 L19641.06 L19641.MD
L19641.03EA 148 EUR
L19641.03 L19641.06 L19641.MD
5-Chinolincarbaldehyd ≥97%
5-Chinolincarbaldehyd

Purity: 97+% CAS No.: 22934-41-4 Molecular Formula: C10H7NO Formula Weight: 157.17 Melting Point: 94-98° Beilstein Reference: 113095

Formel: C₁₀H₇NO
Molecular Weight: 157,17 g/mol
Schmelzpunkt: 94…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00805835
CAS-Nummer: 22934-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR