8-Chinolincarbaldehyd ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
8-Chinolincarboxaldehyd
L19642.03
L19642.06
L19642.03EA
143
EUR
L19642.03
L19642.06
8-Chinolincarbaldehyd ≥98%
8-Chinolincarbaldehyd
Purity: 98+%
CAS No.: 38707-70-9
Molecular Formula: C10H7NO
Formula Weight: 157.17
Melting Point: 92-94°
Beilstein Reference: 113039
Formel:
C₁₀H₇NO Molecular Weight: 157,17 g/mol Schmelzpunkt: 92…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00805837 CAS-Nummer: 38707-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...