2-Chinolincarbonitril ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Quinoline-2-carbonitrile

L19427.03 L19427.06
L19427.03EA 37.2 EUR
L19427.03 L19427.06
2-Chinolincarbonitril ≥97%
2-Chinolincarbonitril

Purity: 97% CAS No.: 1436-43-7 Molecular Formula: C10H6N2 Formula Weight: 154.17 Odour: Odourless Melting Point: 93-96° Boiling Point: 160°/23mm Beilstein Reference: 115213 MDL No.: MFCD00134341

Formel: C₁₀H₆N₂
Molecular Weight: 154,17 g/mol
Siedepunkt: 160 °C (23 mmHg)
Schmelzpunkt: 93…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00134341
CAS-Nummer: 1436-43-7
EINECS: 215-865-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR