4-Chinolincarbonsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Chinolin-4-carbonsäure

L14611.03 L14611.06
L14611.03EA 71.2 EUR
L14611.03 L14611.06
4-Chinolincarbonsäure ≥98%
4-Chinolincarbonsäure

Purity: 98+% CAS No.: 486-74-8 Molecular Formula: C10H7NO2 Formula Weight: 173.17 Melting Point: 253-255° Beilstein Reference: 5224

Formel: C₁₀H₇NO₂
Molecular Weight: 173.17 g/mol
Schmelzpunkt: 253…255 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006782
CAS-Nummer: 486-74-8
EINECS: 207-640-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR