Cloxiquine 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Chlor-8-chinolinol

B22856.18 B22856.30
B22856.18EA 29 EUR
B22856.18 B22856.30
Cloxiquine 95%
Cloxiquine

Purity: 95% CAS No.: 130-16-5 Molecular Formula: C9H6ClNO Formula Weight: 179.61 Melting Point: 122-124° Beilstein Reference: 5289 Merck Reference: 14,2422 MDL No.: MFCD00006788

Formel: C₉H₆ClNO
Molecular Weight: 179.61 g/mol
Siedepunkt: 349
Schmelzpunkt: 122…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006788
CAS-Nummer: 130-16-5
EINECS: 204-978-1
Merck Index: 12,02483

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR