3-Aminochinolin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Chinolin-3-ylamin

A16592.03 A16592.06 A16592.14 AB113632.0025
A16592.03EA 24.9 EUR
A16592.03 A16592.06 A16592.14 ABCRAB113632.0025
3-Aminochinolin 98%
3-Aminochinolin

Purity: 98% CAS No.: 580-17-6 Molecular Formula: C9H8N2 Formula Weight: 144.18 Melting Point: 92-96° Beilstein Reference: 113317

Formel: C₉H₈N₂
Molecular Weight: 144.18 g/mol
Siedepunkt: 311
Schmelzpunkt: 92…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006772
CAS-Nummer: 580-17-6
EINECS: 209-455-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR