5-Aminochinolin 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Chinolin-5-ylamin

L19628.03 L19628.06
L19628.03EA 63.4 EUR
L19628.03 L19628.06
5-Aminochinolin 99%
5-Aminochinolin

Purity: 99% CAS No.: 611-34-7 Molecular Formula: C9H8N2 Formula Weight: 144.18 Melting Point: 107-109° Boiling Point: 310° MDL No.: MFCD00006797

Formel: C₉H₈N₂
Molecular Weight: 144.18 g/mol
Siedepunkt: 310 °C
Schmelzpunkt: 107…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006797
CAS-Nummer: 611-34-7
EINECS: 210-266-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR