6-Aminochinolin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Chinolin-6-ylamin
369640010
369640050
369640250
ACRO369640010EA
34.6
EUR
ACRO369640010
ACRO369640050
ACRO369640250
6-Aminochinolin 98%
6-Aminochinolin
Formel:
C₉H₈N₂ Molecular Weight: 144,18 g/mol Siedepunkt: 192…195 °C (14 mmHg) Schmelzpunkt: 115…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006803 CAS-Nummer: 580-15-4 EINECS: 209-453-0 |
Spezifikation Testergebnisse
Appearance | Yellow to grey-green to brown Powder |
Infrared spectrum | Conforms |
Melting point | 115 to 119 °C |
HPLC | ≥97.5 % |
Titration with HClO4 | 97.5 to 102.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...