6-Aminochinolin 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Chinolin-6-ylamin
L13819.03
L13819.06
L13819.14
L13819.03EA
44.3
EUR
L13819.03
L13819.06
L13819.14
6-Aminochinolin 98%
6-Aminochinolin
Purity: 98%
CAS No.: 580-15-4
Molecular Formula: C9H8N2
Formula Weight: 144.18
Odour: Odourless
Melting Point: 115-119°
Boiling Point: 146°/0.3mm
Beilstein Reference: 113320
MDL No.: MFCD00006803
Formel:
C₉H₈N₂ Molecular Weight: 144.18 g/mol Siedepunkt: 146 °C (0.3 mmHg) Schmelzpunkt: 115…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006803 CAS-Nummer: 580-15-4 EINECS: 209-453-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...