4-Chinolinol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A15859.03 A15859.06 A15859.09
A15859.03EA 28.2 EUR
A15859.03 A15859.06 A15859.09
4-Chinolinol ≥98%
4-Chinolinol

Purity: 98% CAS No.: 611-36-9 Molecular Formula: C9H7NO Formula Weight: 145.16 Melting Point: 201-205° Beilstein Reference: 1524969

Formel: C₉H₇NO
Schmelzpunkt: 201…205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006777
CAS-Nummer: 611-36-9
EINECS: 210-268-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR