8-Chinolinol ACS
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
8-Hydroxyquinoline
121901000
121905000
121900010
121900025
121900250
ACRO121901000EA
56.7
EUR
ACRO121901000
ACRO121905000
ACRO121900010
ACRO121900025
ACRO121900250
8-Chinolinol ACS
8-Chinolinol
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol Siedepunkt: 267 °C (752 mmHg) Schmelzpunkt: 72…74 °C Dichte: 1,03 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006807 CAS-Nummer: 148-24-3 EINECS: 205-711-1 UN: 2811 ADR: 6.1,III Merck Index: 13,04869 |
Spezifikation Testergebnisse
Appearance | White to light yellow to light beige Crystalline powder or crystals or flakes |
Infrared spectrum | Conforms |
Melting point | 72.5 to 74.0 °C |
Residue after ignition | ≤0.05 % |
Suitability | (for Mg determination) Passes test |
Sulfate (SO4) | ≤0.02 % |
Insoluble matter | ≤0.05 % (in alcohol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...