2,8-Chinolindiol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2,8-Dihydroxyquinoline

37640-1G-F 37640-5G-F
SIAL37640-1G-FEA 37.4 EUR
SIAL37640-1G-F SIAL37640-5G-F
2,8-Chinolindiol, Sigma-Aldrich®
2,8-Chinolindiol
Formel: C₉H₇NO₂
Molecular Weight: 161.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00216696
CAS-Nummer: 15450-76-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR