6-Hydroxychinaldin

Lieferant: CHEMPUR
Achtung

Synonyme: 2-Methyl-6-chinolinol

BTB12328.1 BTB12328.10
CHMPBTB12328.1EA 0 EUR
CHMPBTB12328.1 CHMPBTB12328.10
6-Hydroxychinaldin
6-Hydroxychinaldin
Formel: C₁₀H₉NO
Molecular Weight: 159,19 g/mol
MDL: MFCD00047600
CAS-Nummer: 613-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR