4-Methylchinolin ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Lepidin

A14040.09 A14040.18 A14040.30
A14040.09EA 36.2 EUR
A14040.09 A14040.18 A14040.30
4-Methylchinolin ≥97%
4-Methylchinolin

Purity: 98% CAS No.: 491-35-0 Molecular Formula: C10H9N Formula Weight: 143.19 Flash Point: 110°(230°F) Refractive Index: 1.6200 Melting Point: 9-10° Boiling Point: 260-263° Beilstein Reference: 110926 Merck Reference: 14,5441 MDL No.: MFCD00006784

Formel: C₁₀H₉N
Molecular Weight: 143,19 g/mol
Siedepunkt: 260…263 °C
Schmelzpunkt: 9…10 °C
Dichte: 1,083
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006784
CAS-Nummer: 491-35-0
EINECS: 207-734-2
Merck Index: 14,5441

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR