8-Methylchinolin ≥97%

Lieferant: Thermo Scientific
Achtung

L03288.06 L03288.14
L03288.06EA 42.7 EUR
L03288.06 L03288.14
8-Methylchinolin ≥97%
8-Methylquinoline

Purity: 97+% CAS No.: 611-32-5 Molecular Formula: C10H9N Formula Weight: 143.19 Flash Point: 105°(221°F) Refractive Index: 1.6150 Odour: Unpleasant Melting Point: -80° Boiling Point: 134-136°/25mm Beilstein Reference: 111340 MDL No.: MFCD00006810

Formel: C₁₀H₉N
Molecular Weight: 143,19 g/mol
Siedepunkt: 134…136 °C (25 mmHg)
Schmelzpunkt: –80 °C
Dichte: 1,074
Flammpunkt: 105 °C (221 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006810
CAS-Nummer: 611-32-5
EINECS: 210-264-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR