Bromcyclobutan 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Cyclobutylbromid
A12817.14
A12817.03
A12817.06
AB109056.0025
A12817.14EA
687
EUR
A12817.14
A12817.03
A12817.06
ABCRAB109056.0025
Bromcyclobutan 95%
Bromcyclobutan
Purity: 95%
CAS No.: 4399-47-7
Molecular Formula: C4H7Br
Formula Weight: 135.01
Flash Point: 22°(71°F)
Refractive Index: 1.4795
Boiling Point: 107-108°
Beilstein Reference: 1900415
UN No.: 1993
MDL No.: MFCD00001317
Formel:
C₄H₇Br Molecular Weight: 135 g/mol Siedepunkt: 107…108 °C Dichte: 1,431 Flammpunkt: 22 °C (71 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001317 CAS-Nummer: 4399-47-7 EINECS: 224-530-9 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...