Bromcyclopentan

Lieferant: FLUOROCHEM

Synonyme: Cyclopentylbromid

021866-1KG 021866-250G
FLUO021866-1KGEA 233 EUR
FLUO021866-1KG FLUO021866-250G
Bromcyclopentan
Bromcyclopentan
Formel: C₅H₉Br
Molecular Weight: 149.03 g/mol
Siedepunkt: 138
Dichte: 1.39 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001359
CAS-Nummer: 137-43-9
EINECS: 205-294-6
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR