Bromcyclopropan ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Cyclopropylbromid

A14996.09 A14996.18 A14996.30
A14996.09EA 69.6 EUR
A14996.09 A14996.18 A14996.30
Bromcyclopropan ≥99%
Bromcyclopropan

Purity: 99% CAS No.: 4333-56-6 Molecular Formula: C3H5Br Formula Weight: 120.98 Storage Temperature: 2°C to 8°C Flash Point: -6°(21°F) Refractive Index: 1.4600 Odour: Recognizable Boiling Point: 68-70° Beilstein Reference: 1900287 UN No.: 1993 MDL No.: MFCD00001271

Formel: C₃H₅Br
Molecular Weight: 120,98 g/mol
Siedepunkt: 68…70 °C
Dichte: 1,515
Flammpunkt: –6 °C (21 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00001271
CAS-Nummer: 4333-56-6
EINECS: 224-375-7
UN: 1993
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR