Cycloheptanon ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Suberon
A13421.18
A13421.30
A13421.18EA
44.3
EUR
A13421.18
A13421.30
Cycloheptanon ≥98%
Cycloheptanon
Purity: 99%
CAS No.: 502-42-1
Molecular Formula: C7H12O
Formula Weight: 112.17
Flash Point: 55°(131°F)
Refractive Index: 1.4610
Odour: Sweet
Melting Point: -21°
Boiling Point: 179-181°
Beilstein Reference: 969823
Merck Reference: 14,2722
UN No.: 1224
MDL No.: MFCD00004159
Formel:
C₇H₁₂O Molecular Weight: 112,17 g/mol Siedepunkt: 179…181 °C Schmelzpunkt: –21 °C Dichte: 0,956 Flammpunkt: 55 °C (131 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004159 CAS-Nummer: 502-42-1 EINECS: 207-937-6 UN: 1224 ADR: 3,III Merck Index: 14,2722 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...