trans-1,2-Cyclobutandicarbonsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

28684-1G 28684-5G
SIAL28684-1GEA 196 EUR
SIAL28684-1G SIAL28684-5G
trans-1,2-Cyclobutandicarbonsäure, Sigma-Aldrich®
trans-1,2-Cyclobutandicarbonsäure
Formel: C₆H₈O₄
Molecular Weight: 144,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00068260
CAS-Nummer: 1124-13-6
EINECS: 214-392-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR