Chlorcyclobutan
Lieferant: FLUOROCHEM
Synonyme:
Cyclobutylchlorid
003369-1G
003369-5G
FLUO003369-1GEA
116
EUR
FLUO003369-1G
FLUO003369-5G
Chlorcyclobutan
Chlorcyclobutan
Formel:
C₄H₇Cl Molecular Weight: 90,55 g/mol Siedepunkt: 80,2 °C (1013 hPa) Dichte: 0,991 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00013736 CAS-Nummer: 1120-57-6 EINECS: 214-314-2 UN: 1127 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...