Chlorcyclopentan ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Cyclopentyl chloride
B25023.22
B25023.36
B25023.22EA
49.3
EUR
B25023.22
B25023.36
Chlorcyclopentan ≥99%
Chlorcyclopentan
Purity: 99%
CAS No.: 930-28-9
Molecular Formula: C5H9Cl
Formula Weight: 104.58
Flash Point: 15°(59°F)
Refractive Index: 1.4512
Melting Point: -94°
Boiling Point: 113-114°
Beilstein Reference: 1900557
UN No.: 1993
MDL No.: MFCD00001360
Formel:
C₅H₉Cl Molecular Weight: 104.58 g/mol Siedepunkt: 113…114 °C Schmelzpunkt: –94 °C Dichte: 1.003 Flammpunkt: 15 °C (59 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001360 CAS-Nummer: 930-28-9 EINECS: 213-212-5 UN: 1993 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...