Cyclohexanol ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Hydroxycyclohexan, 1-Hydroxycyclohexan
A17576.0F
A17576.AE
A17576.AP
A17576.0FEA
82.9
EUR
A17576.0F
A17576.AE
A17576.AP
Cyclohexanol ≥99%
Cyclohexanol
Purity: 99%
CAS No.: 108-93-0
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: 67°(152°F)
Physical Form: Liquid/low-melting solid
Density: 0.951
Refractive Index: 1.4650
Appearance: Colourless
Melting Point: 23-24°
Boiling Point: 160-161°
Beilstein Reference: 906744
Merck Reference: 14,2725
MDL No.: MFCD00003855
Formel:
C₆H₁₂O Molecular Weight: 100,16 g/mol Siedepunkt: 160…161 °C Schmelzpunkt: 23…24 °C Dichte: 0,951 Flammpunkt: 67 °C (152 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003855 CAS-Nummer: 108-93-0 EINECS: 203-630-6 Merck Index: 14,2725 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...