Cyclopentanthiol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Mercaptocyclopentan, Cyclopentylmercaptan

L03577.06 L03577.14
L03577.06EA 36 EUR
L03577.06 L03577.14
Cyclopentanthiol ≥97%
Cyclopentanthiol

Purity: 97% CAS No.: 1679-07-8 Molecular Formula: C5H10S Formula Weight: 102.20 Flash Point: 25°(77°F) Refractive Index: 1.4900 Odour: Stench Boiling Point: 131-132° Beilstein Reference: 2343861 UN No.: 3336 MDL No.: MFCD00001369

Formel: C₅H₁₀S
Molecular Weight: 102.2 g/mol
Siedepunkt: 131…132 °C
Dichte: 0.957
Flammpunkt: 25 °C (77 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001369
CAS-Nummer: 1679-07-8
EINECS: 216-841-3
UN: 3336
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR