(3-Chlorpropyl)dimethylammoniumchlorid 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Dimethylaminopropylchloride hydrochloride, 3-Dimethylaminopropyl chloride hydrochloride, N-(3-Chloropropyl)-N,N-dimethylammonium chloride, 3-Chloro-N,N-dimethylpropan-1-amine hydrochloride

115911000 115915000
ACRO115911000EA 29.1 EUR
ACRO115911000 ACRO115915000
(3-Chlorpropyl)dimethylammoniumchlorid 99%
3-Chloropropyldimethylammonium chloride
Formel: C₅H₁₃Cl₂N
Molecular Weight: 158,07 g/mol
Schmelzpunkt: 139…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012521
CAS-Nummer: 5407-04-5
EINECS: 226-467-2

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (105°C, 3 hrs)
Titration with HClO4 98.5 to 101.5 % (on dry substance)
Chloride (Cl) 21.9 to 22.9 % (titr.)
Chlorine (Cl) 44 to 45.5 % (total)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR