Tropin 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol

140560250 140561000
ACRO140560250EA 69.3 EUR
ACRO140560250 ACRO140561000
Tropin 98%
Tropin
Formel: C₈H₁₅NO
Molecular Weight: 141.21 g/mol
Siedepunkt: 233 °C (760 mmHg)
Schmelzpunkt: 62…64 °C
Dichte: 1.04 g/cm³
Lagertemperatur: Kühlschrank
MDL: MFCD00180667
CAS-Nummer: 120-29-6
EINECS: 204-384-2

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤2 % (K.F.)
Titration with HClO4 ≥97.5 % (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR