alle-trans-Retinol 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: all-trans-Retinol, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-ol, Vitamin A alcohol

347071000 347070010 347072500 347075000
ACRO347071000EA 55.6 EUR
ACRO347071000 ACRO347070010 ACRO347072500 ACRO347075000
alle-trans-Retinol 95%
alle-trans-Retinol
Formel: C₂₀H₃₀O
Molecular Weight: 286,46 g/mol
Schmelzpunkt: 61…63 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00001552
CAS-Nummer: 68-26-8
EINECS: 200-683-7

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to orange Powder
Infrared spectrum Conforms
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR