Brij® 98
Lieferant: Thermo Scientific
|
Achtung
|
347185000
347181000
ACRO347185000EA
83.8
EUR
ACRO347185000
ACRO347181000
Brij® 98
Brij® 98
Formel:
C₅₈H₁₁₆O₂₁ Molecular Weight: 1149,55 g/mol Siedepunkt: >100 °C (760 mmHg) Schmelzpunkt: 30 °C Dichte: 1,07 g/cm³ Flammpunkt: >150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00074369 CAS-Nummer: 9004-98-2 EINECS: 500-016-2 |
Spezifikation Testergebnisse
Appearance | White to cream Waxy solid |
Infrared spectrum | Conforms |
Water | ≤3 % |
Additional info | remark: without antioxidant |
Impurity | ≤1 ppm (Free ethyleneoxide) |
Dioxane | ≤10 ppm |
Acid value | ≤1 mg KOH/g |
Hydroxyl value | 50 to 65 mg KOH/g |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...