1-(4-(Trifluormethyl)phenyl)piperazin 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: N-(α,α,α-Trifluoro-p-tolyl)piperazine

367120010 367120050
ACRO367120010EA 46.3 EUR
ACRO367120010 ACRO367120050
1-(4-(Trifluormethyl)phenyl)piperazin 98%
1-(4-(Trifluormethyl)phenyl)piperazin
Formel: C₁₁H₁₃F₃N₂
Molecular Weight: 230,23 g/mol
Schmelzpunkt: 89…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040765
CAS-Nummer: 30459-17-7
EINECS: 250-210-3
UN: 3259
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to light yellow Crystals
Infrared spectrum Conforms
Melting point 85 to 90 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR