1-Boc-4-hydroxypiperidin 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-Boc-4-piperidinol, tert-butyl 4-hydroxypiperidine-1-carboxylate, N-BOC-4-Hydroxypiperidine, tert-Butyl 4-hydroxypiperidine-1-carboxylate

368270050 368270250 368271000
ACRO368270050EA 56.3 EUR
ACRO368270050 ACRO368270250 ACRO368271000
1-Boc-4-hydroxypiperidin 97%
1-Boc-4-hydroxypiperidin
Formel: C₁₀H₁₉NO₃
Molecular Weight: 201.27 g/mol
Schmelzpunkt: 60…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01075174
CAS-Nummer: 109384-19-2

Bestellen

Spezifikation Testergebnisse

Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 60 to 65 °C
GC ≥96.0 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR