Indium(III)trifluormethansulfonat 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Indium(III) trifluoromethanesulphonate, Indium triflate
368540050
368540250
ACRO368540050EA
103
EUR
ACRO368540050
ACRO368540250
Indium(III)trifluormethansulfonat 98%
Indium(III)trifluormethansulfonat
Formel:
C₃F₉InO₉S₃ Molecular Weight: 562,03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144478 CAS-Nummer: 128008-30-0 UN: 3261 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | White to almost white Powder and/or granules |
Infrared spectrum | Conforms |
Titration Complexometric | 19.9 to 20.9 % (In) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...