Indium(III)trifluormethansulfonat ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Indium(III) trifluoromethanesulphonate, Indium triflate
40131.09
40131.18
40131.09EA
183
EUR
40131.09
40131.18
Indium(III)trifluormethansulfonat ≥99%
Indium(III)trifluormethansulfonat
Purity: 99% min
CAS No.: 128008-30-0
Molecular Formula: In(CF3SO3)3
Formula Weight: 562.02
Physical Form: Powder
Sensitivity: Hygroscopic
UN No.: 3261
MDL No.: MFCD00144478
Formel:
C₃F₉InO₉S₃ Schmelzpunkt: Zersetzung Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144478 CAS-Nummer: 128008-30-0 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...