Thulium(III)trifluormethansulfonat ≥98%

Lieferant: Thermo Scientific
Achtung

40313.04 40313.09
40313.04EA 109 EUR
40313.04 40313.09
Thulium(III)trifluormethansulfonat ≥98%
Thulium(III)trifluormethansulfonat

Purity: 98% CAS No.: 141478-68-4 Molecular Formula: C3F9O9S3Tm Formula Weight: 616.13 Physical Form: Powder

Formel: C₃F₉O₉S₃Tm·H₂O
Molecular Weight: 616,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00209617
CAS-Nummer: 141478-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR