Scandium(III)trifluormethansulfonat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Scandium(III) trifluoromethanesulphonate, Scandium triflate
40566.03
40566.06
40566.MD
40566.03EA
93.3
EUR
40566.03
40566.06
40566.MD
Scandium(III)trifluormethansulfonat ≥98%
Scandium(III)trifluormethansulfonat
Purity: 98%
CAS No.: 144026-79-9
Molecular Formula: Sc(OSO2CF3)3
Formula Weight: 492.16
Physical Form: Powder
Sensitivity: Hygroscopic
MDL No.: MFCD00798539
Formel:
C₃F₉O₉S₃Sc Molecular Weight: 492.17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192433 CAS-Nummer: 144026-79-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...