(Triphenylphosphin)gold(I)chlorid ≥99,99% (Metall-Basis) Au ≥ 39.3%, Premion®

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Chlorotriphenylphosphine gold(I), Chloro(triphenylphosphine)gold(I), Triphenylphosphinechlorogold

41007.02 41007.03 41007.06
41007.02EA 200 EUR
41007.02 41007.03 41007.06
(Triphenylphosphin)gold(I)chlorid ≥99,99% (Metall-Basis) Au ≥ 39.3%, Premion®
(Triphenylphosphine)gold(I) chloride

Purity: 99.99% CAS No.: 14243-64-2 Molecular Formula: AuCl[P(C6H5)3] Formula Weight: 494.71 Physical Form: Powder Odour: Odourless MDL No.: MFCD00009588

Formel: C₁₈H₁₅AuClP
Molecular Weight: 494.71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009588
CAS-Nummer: 14243-64-2
EINECS: 238-117-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR