trans-Bis(triphenylphosphin)palladium(II)chlorid ≥99,95% (Metall-Basis) Pd ≥ 14.7%, Premion®
Lieferant: Thermo Scientific
Synonyme: trans-Dichlorobis(triphenylphosphine)palladium(II)
41245.03 41245.06 41245.14
41245.03EA 71.7 EUR
41245.03 41245.06 41245.14
trans-Bis(triphenylphosphin)palladium(II)chlorid ≥99,95% (Metall-Basis) Pd ≥ 14.7%, Premion®
trans-Bis(triphenylphosphin)palladium(II)chlorid
Purity: 99.95% CAS No.: 13965-03-2 Molecular Formula: PdCl2[P(C6H5)3]2 Formula Weight: 701.91 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Yellow Odour: Odourless Melting Point: ca 310° dec. MDL No.: MFCD00009593
Formel: C₃₆H₃₀Cl₂P₂Pd
Molecular Weight: 701,91 g/mol
Schmelzpunkt: ∼ 310 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009593
CAS-Nummer: 13965-03-2
EINECS: 237-744-2