Aluminiumtriethanolat
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Aluminiumethoxid, Aluminiumtriethylat
41336.09
41336.18
41336.30
41336.09EA
51.8
EUR
41336.09
41336.18
41336.30
Aluminiumtriethanolat
Aluminiumtriethanolat
CAS No.: 555-75-9
Molecular Formula: C6H15AlO3
Formula Weight: 162.16
Physical Form: Powder/ Chunks
Density: 1.142
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 154-159°
Boiling Point: 320°
Merck Reference: 14,338
UN No.: 2925
MDL No.: MFCD00009658
Formel:
C₆H₁₅AlO₃ Molecular Weight: 162,16 g/mol Siedepunkt: 320 °C Schmelzpunkt: 154…159 °C Dichte: 1,142 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009658 CAS-Nummer: 555-75-9 EINECS: 209-105-8 ADR: 4.1,II Merck Index: 14,338 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...