1-(2-Aminoethyl)piperazin 98%
Lieferant: Thermo Scientific
|
Gefahr
|
A10154.0B
A10154.30
A10154.0BEA
76.9
EUR
A10154.0B
A10154.30
1-(2-Aminoethyl)piperazin 98%
1-(2-Aminoethyl)piperazin
Purity: 98%
CAS No.: 140-31-8
Molecular Formula: C6H15N3
Formula Weight: 129.21
Flash Point: 93°(199°F)
Refractive Index: 1.5000
Melting Point: -19°
Boiling Point: 222°
Beilstein Reference: 104363
UN No.: 2815
MDL No.: MFCD00005971
Formel:
C₆H₁₅N₃ Molecular Weight: 129,21 g/mol Siedepunkt: 222 °C Schmelzpunkt: –19 °C Dichte: 0,983 Flammpunkt: 93 °C (199 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005971 CAS-Nummer: 140-31-8 EINECS: 205-411-0 UN: 2815 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...