Isonicotinaldehyd ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Pyridine-4-carboxaldehyde, 4-Pyridinecarboxaldehyde, 4-Pyridinecarbaldehyde

A10656.22 A10656.14 A10656.36 AB121167.0025 AB121167.0100
A10656.22EA 102 EUR
A10656.22 A10656.14 A10656.36 ABCRAB121167.0025 ABCRAB121167.0100
Isonicotinaldehyd ≥97%
Isonicotinaldehyd

Purity: 97% CAS No.: 872-85-5 Molecular Formula: C6H5NO Formula Weight: 107.11 Storage Temperature: 2°C to 8°C Flash Point: 75°(167°F) Refractive Index: 1.5440 Melting Point: -4° Boiling Point: 198° Beilstein Reference: 105342 MDL No.: MFCD00006425

Formel: C₆H₅NO
Molecular Weight: 107,11 g/mol
Siedepunkt: 198 °C
Schmelzpunkt: –4 °C
Dichte: 1,132
Flammpunkt: 75 °C (167 °F)
MDL: MFCD00006425
CAS-Nummer: 872-85-5
EINECS: 212-832-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR