Tetrabutylammoniumtrifluormethansulfonat ≥99%

Lieferant: Thermo Scientific
Achtung

A11013.06 A11013.14 A11013.22
A11013.06EA 43.3 EUR
A11013.06 A11013.14 A11013.22
Tetrabutylammoniumtrifluormethansulfonat ≥99%
Tetrabutylammonium trifluoromethanesulphonate

Purity: 99% CAS No.: 35895-70-6 Molecular Formula: [CH3(CH2)3]4NCF3SO3 Formula Weight: 391.54 Melting Point: 112-115° Beilstein Reference: 4172978

Formel: C₁₇H₃₆F₃NO₃S
Molecular Weight: 391,54 g/mol
Schmelzpunkt: 112…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042585
CAS-Nummer: 35895-70-6
EINECS: 252-783-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR